Learn More
Med Vet International McKesson Steam Indicator Tape, 1 Inch x 60 Yard, 73-BT36, Each
Supplier:  Med Vet International 524891RL
Encompass_Preferred
Steam Indicator Tape McKesson 1 Inch X 60 Yard Steam Blue Blue steam tape has a more aggressive adhesive and is for disposable wraps only For use in securing wrapped instrument sets before, during and after sterilization Each roll of McKesson tape is individually packaged to ensure quality at time of use 1 inch x 60 yards
Catalog No. 50-304-4637