Learn More
Med Vet International MedFix Water Resistant Dressing Retention Tape with Liner, White, 6 Inch X 11 Yard, Nonwoven, NonSterile, MSC4006, 1/Box
Supplier:  Med Vet International 541968BX
Encompass_Preferred
For flexibility and customized sizing Low sensitivity adhesive, gentle for the patient Medfix has a printed s-c release liner Water resistant Indications: to secure primary dressings and to secure gastrostomy tubes and other feeding tubes Contraindicated as a primary dressing May be left in place for up to 7 days
Catalog No. 50-304-4249