Learn More
Med Vet International Perforated Medical Tape 3M Medipore H White 2 inch X 10 Yard Soft Cloth Nonsterile, 2862, 12/Case
Supplier:  Med Vet International 324081CS
Encompass_Preferred
Water-resistant, non-sterile 3M Medipore H tape is suitable for securing dressings, tubing and catheters The multi-directional stretch white fabric allows for freedom of movement and wound swelling 2-inch x 10-yard 3M Medipore H soft cloth tape roll includes perforations for quick tearing by hand Porous, breathable cloth material allows for aeration while remaining gentle on the skin
Catalog No. 50-283-1527