Learn More
Grainger Tape Purple 50 ft L 4 in W
Supplier:  Grainger 14A586
Encompass_Preferred
The Grainger 14A586 is a Westward 1/4-inch drive socket set that includes both standard and deep sockets ranging from 4mm to 14mm. The set has a 72 tooth ratchet with a 5 degree arc swing and a knurled handle. The sockets are made from chrome vanadium steel and have a black phosphate finish. The set comes packaged in a molded case for storage and portability. With both standard and deep sockets, this socket set is versatile for automotive and machinery applications.
Catalog No. 50-275-3627