Learn More
Med Vet International Icu Medical IV Administration Set, 80in., Dehp Free, Each
Supplier:  Med Vet International 1267228
Encompass_Preferred
LifeShield primary set with convertible piercing pin and sight chamber Slide clamp CAIR clamp Prepierced Y-site Option-Lok male adapter 80" / 203.2 cm 15 drops/mL
Catalog No. 50-282-9041