Learn More
MSC DISP CHEM RESIST CT APRN
Supplier:  MSC 86529963
Encompass
Disposable & Chemical Resistant Aprons; Garment Style: Coat Apron ; Garment Type: Dual Hazard ; Garment Length (Inch): 83 ; Garment Width (Inch): 55 ; Material: Multi-Layer Non-Woven Barrier Laminate Fabric ; Closure Type: Ties In Back; Hook & Loop
Catalog No. 50-273-6685