Learn More
Essendant TAPEELECTRICAL 88 3/4X66
Supplier:  Essendant MMM06143
Encompass_Preferred
Polyvinyl chloride (PVC) backing Pressure sensitive rubber based adhesive Compatible with solid dielectric cable insulations Compatible with rubber and synthetic splicing compounds as well as epoxy and polyurethane resins Inhibits corrosion of electrical conductors Dispenser Included- No Adhesive Material- Natural Rubber Backing Material- Vinyl Tensile Strength- 20 lb/in
Catalog No. 50-000-33377