Learn More
Essendant TAPEFLATBACK 60YD BOXED
Supplier:  Essendant MMM25634
Encompass_Preferred
Scotch Printable Flatback Paper Tape can be used for masking as well as general holding bundling and splicing applications however it s specifically designed for operations requiring colored tapes for marking or identification It adheres best to paper film fabric and yarn The tape s backing surface may be easily marked and written on and it works well in many graphic arts applications The heavyweight thick colored flatback paper backing tears by hand conforms to corners curves and irregular surfaces and removes without slivering making the tape very easy to manipulate and apply Core Size- 3 Dispenser Included- No Adhesive Material- Natural Rubber Backing Material- Paper
Catalog No. 50-000-33083