Learn More
Essendant TAPELABELELEC1BK/WHT
Supplier:  Essendant DYM53713
Encompass_Preferred
Designed to work with LabelManager and LabelWriter Duo label makers DYMO D1 labels are both high quality and versatile enough to handle most labeling tasks Available in multiple color combinations and widths this label maker tape is a great way to get organized The label cassettes also print using thermal transfer technology which prints without expensive ink or toner Continuous Label-Maker Tape Cassette Perfect for many tasks around the home or office can be removed without leaving adhesive residue Label Size - text- 1x23 ft Label Color(s)- White Machine Compatibility- DYMO LabelManager DYMO LabelPoint DYMO LabelWriter Duo DYMO Rhino Older DYMO Label Printers That Use D1 Tapes Text/Label Colors- Black Print on White Tape
Catalog No. 50-000-35646