Learn More
Glass Expansion Tapered Quartz Injector 1.8mm
SDP
Supplier:  Glass Expansion 318083576
Encompass_Preferred
Quartz Tapered Injector with 1.8mm ID. A 1.8mm ID quartz tapered injector suitable for use with aqueous and less volatile organic solvent-based matrices. Compatible with Agilent ICP-OES 5100/5110/5800/5900 D-Torch (30-808-3560). Not suitable for use with HF. When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7232