Learn More
MSC GF Gage Plug Thread Gage, 1-1/4-7, 3B, Double End, 1 ea
Supplier: MSC 55311773
This double end plug thread gage is designed for precision measurement of 1-1/4-7 threads. It is manufactured from hardened tool steel and includes a handle for ease of use in industrial quality control applications.
- Features a 3B class classification
- Double end configuration includes both go and no go gauging
- Go pitch diameter is 1.1572 in.
- No go pitch diameter is 1.1644 in.
- Includes handle