Learn More
Essendant TAPESTRAPPING2X360
Supplier:  Essendant MMM50
Encompass_Preferred
Provides very good holding power under a wide range of conditions Scotch Reinforced Strength Shipping and Strapping Tape combines a strong polypropylene backing reinforced with continuous glass yarn filaments and a synthetic rubber resin adhesive for long-term holding power to most fiberboard surfaces including recycled and printed The adhesive provides good initial adhesion and holds well with minimal rub-down The tough clear film backing is abrasion- and moisture-resistant Features a 320 lbs pulling strength rating Grade- Heavy-Duty Core Size- 1 5 Dispenser Included- Yes Adhesive Material- Hot Melt Synthetic Rubber
Catalog No. 50-000-31764