Learn More
Abcam TAPI-0 (TNF alpha processing inhibitor-0), Matri x metalloprotease (MMP) and TACE inhibitor, 1MG
Supplier:  Abcam AB1414971MG
Encompass_Preferred
MW 456.5 Da, Purity >95%. Inhibits TNF-α processing. Also a general matrix metalloprotease and TACE inhibitor.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6564