Learn More
Abcam TAPI-1, TNF alpha processing inhibitor, 1MG
Supplier:  Abcam AB1422181MG
Encompass_Preferred
MW 499.6 Da, Purity >95%. Stable, selective inhibitor of TNF-α processing (IC₅₀ = ~100 nM). Inhibits TACE (P< 0.01) and other metalloproteases in vivo. Blocks release of TNF-α (IC₅₀ = 50-100 μM), APP (IC₅₀ = 1-8 μM), ACE (IC₅₀ = 10 μM), IL6R (IC₅₀ = 5-10 μM), TNFRI (IC₅₀ = 5-10 μM), TNFRII (IC₅₀ = 25-50 μM), and ObRa (leptin receptor), but not CSF-1 in vivo. Structural analog of TAPI-0.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6278