Learn More
Selleck Chemical LLC Tapinarof 5mg
SDP
Supplier:  Selleck Chemical LLC S97005MG
Encompass_Preferred
Tapinarof (GSK2894512, Benvitimod, WBI 1001, DHPS, DMVT 505) is a natural agonist ofaryl hydrocarbon receptor (AhR) and induces nuclear translocation of AhR in immortalized keratinocytes (HaCaT) with EC50 of 0.16 nM. Tapinarof induces cellularapoptosis in CD4+ T cells in a dosedependent manner with IC50 of 5.2 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3498