Learn More
Medchemexpress LLC Taprenepag | 752187-80-7 | 98.8% | 478.52 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1489950MG
Encompass_Preferred
Taprenepag (CP-544326) is a potent and selective prostaglandin EP(2) agonist with IC50s of 10 and 15 nM for human and rat EP2, respectively. It demonstrates selectivity for EP2 over other EP receptors (IC50s > 3200 nM for EP1, EP3, and EP4) and a panel of 37 G protein-coupled receptors. It increases cAMP levels in HEK293 cells expressing human EP2 (EC50=2.8 nM) and has poor corneal permeability in an ex vivo rabbit corneal model.
  • Potent and selective prostaglandin EP(2) agonist
  • Shows selectivity for EP2 over other EP receptors
  • Increases cAMP levels in HEK293 cells
  • Has poor corneal permeability in an ex vivo rabbit corneal model
Catalog No. 50-003-61160