Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Taprenepag | 752187-80-7 | 98.8% | 478.52 g/mol | C24H22N4O5S | 10 MG
SDP

Supplier:  Medchemexpress LLC HY1489910MG

Encompass_Preferred

Taprenepag is a potent, selective prostaglandin EP2 receptor agonist used as a research reagent to study EP2-mediated signaling. It displays low-nanomolar activity in human and rat assays, strong selectivity over other prostaglandin receptors, and is provided as a high-purity material for laboratory applications.

  • Potent EP2 agonist with low-nanomolar activity.
  • High selectivity versus other prostaglandin receptors.
  • High purity suitable for research use (98.8%).
  • Molecular weight 478.52 g/mol and formula C24H22N4O5S.
  • Storage recommendations: -20°C under nitrogen; in solvent -80°C for 6 months or -20°C for 1 month.
  • Supplied in small research quantities; verify package size with supplier.

Catalog No. 50-002-85610


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.