Learn More
Medchemexpress LLC Taprenepag | 752187-80-7 | MFCD18633300 | 98.79% | 478.52 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14899100MG
Encompass_Preferred
Taprenepag (CP-544326) is a potent and selective prostaglandin EP(2) agonist. It demonstrates high selectivity for EP2 over other EP receptors and a panel of 37 G protein-coupled receptors. It increases cAMP levels in HEK293 cells expressing human EP2.
  • Potent and selective prostaglandin EP(2) agonist
  • High selectivity for EP2 over other EP receptors
  • Increases cAMP levels in HEK293 cells expressing human EP2
  • Poor corneal permeability in an ex vivo rabbit corneal model
Catalog No. 50-003-61134