Learn More
Medchemexpress LLC Taranabant (1R,2R)stereoisomer | 701977-08-4 | 98.2% | 515.95 g/mol | C27H25ClF3N3O2 | 1 ML

Supplier: Medchemexpress LLC HY10013B10MM 1ML
Taranabant (1R,2R) stereoisomer is the R-enantiomer of taranabant, a potent and selective cannabinoid 1 (CB1) receptor inverse agonist supplied for research use. It is provided at high purity and is available both as a 10 mM solution in DMSO and in multiple solid sizes for in vitro pharmacology and biochemical studies.
- High purity (98.15%).
- CAS number 701977-08-4.
- Molecular weight 515.95 g/mol.
- Available as 10 mM solution in DMSO and solids (2 mg-500 mg).
- Suitable for in vitro pharmacology and receptor binding studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.