Learn More
Medchemexpress LLC Taraxerone | 514-07-8 | 424.70 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN11771MG
Encompass_Preferred
Taraxerone is isolated from Sedum sarmentosum. It enhances alcohol dehydrogenase (ADH) and acetaldehyde dehydrogenase (ALDH) activities. It also significantly lowers plasma alcohol and acetaldehyde concentrations in mice and prevents the decrease of catalase, superoxide dismutase, and reduced glutathione concentrations induced by ethanol administration.
  • Isolated from Sedum sarmentosum.
  • Enhances alcohol dehydrogenase (ADH) and acetaldehyde dehydrogenase (ALDH) activities.
  • Lowers plasma alcohol and acetaldehyde concentrations.
  • Prevents reduction of catalase, superoxide dismutase, and glutathione due to ethanol.
Catalog No. 50-004-15119