Learn More
Medchemexpress LLC TARC/CCL17 protein, dog (HEK293, His) | >95.0% | 10UG
SDP
Supplier:  Medchemexpress LLC HYP70055210UG
Encompass_Preferred
Recombinant TARC/CCL17 protein derived from dog, expressed in HEK293 cells and C-terminally His-tagged. The product corresponds to residues A24-S99 (accession Q95N01), is supplied lyophilized, and is formulated in PBS with 6% trehalose; predicted molecular mass ~9.4 kDa. Provided with supporting documents and recommended storage for research use only.
  • Expression host HEK293 for mammalian expression
  • C-terminal His tag for purification and detection
  • Residues A24-S99 sequence confirmed
  • Lyophilized formulation in PBS with 6% trehalose
  • Purity >95.0% by reducing SDS-PAGE
  • Supplied with COA, SDS, and data sheet for quality control
Catalog No. 50-001-75470