Learn More
Medchemexpress LLC Tarenflurbil | 51543-40-9 | MFCD00869714 | >98.0% | 244.26 g/mol | C15H13FO2 | 1 ML

Supplier: Medchemexpress LLC HY1029110MM 1ML
Tarenflurbil is the R-enantiomer of flurbiprofen supplied for research use in neuroscience and Alzheimer's disease studies. It modulates amyloid processing and inhibits RXRα LBD binding, making it suitable for in vitro and analytical applications. Typical purity is greater than 98% and molecular weight is 244.26 g/mol.
- R-enantiomer of flurbiprofen
- Used in Alzheimer's disease and neuroscience research
- Molecular formula C15H13FO2; molecular weight 244.26 g/mol
- Purity >98%
- Soluble in DMSO (100 mg/mL) and ethanol (50 mg/mL)
- Store powder at -20°C; store solutions at -80°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.