Learn More
Medchemexpress LLC Taribavirin hydrochloride | 40372-00-7 | MFCD00876899 | 99.9% | 279.68 g/mol | C8H14ClN5O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10545A10MM 1ML
Encompass_Preferred
Taribavirin hydrochloride is the hydrochloride salt form of taribavirin, a ribavirin prodrug used in antiviral research as an inosine monophosphate dehydrogenase (IMPDH) inhibitor. The product is offered as a high-purity solid and as a ready-to-use 10 mM solution in DMSO (1 mL), designed for laboratory studies that require precise dosing and reliable stability.
  • High purity suitable for research-grade experiments.
  • Provided as a 10 mM solution in DMSO for immediate use.
  • Also available as a solid for custom formulation.
  • Stable when stored sealed; solvent storage recommended at -80°C for long term.
  • Molecular weight 279.68 g/mol and CAS 40372-00-7 for identification.
  • Useful as an IMPDH inhibitor and ribavirin prodrug in antiviral studies.
Catalog No. 50-002-40195