Learn More
Medchemexpress LLC TAS-103 dihydrochloride | 174634-09-4 | MFCD00943251 | 98.5% | 406.31 g/mol | C20H21Cl2N3O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY13758A5MG
Encompass_Preferred
TAS-103 dihydrochloride is a small-molecule dual inhibitor of DNA topoisomerase I and II used in cancer research. Supplied as the dihydrochloride salt with reported high purity, it is provided as a powder suitable for biochemical and cellular assays and laboratory research.
Catalog No. 50-002-22016