Learn More
TARGETMOL CHEMICALS INC TAS1553 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T602175MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TAS1553 is a highly effective orally bioavailable protein-protein interaction (PPI) inhibitor with an IC50 value of 0.0396 uM. It effectively hinders DNA replication and diminishes the intracellular dATP pool. Moreover TAS1553 induces apoptosis making it a valuable compound in cancer research [1]. purity: 97%
Catalog No. 50-257-9636