Learn More
TARGETMOL CHEMICALS INC TASIN-1 HYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T248555MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TASIN-1 Hydrochloride (TASIN-1 HCl) is a selective inhibitor of truncated APC that acts by selectively killing colorectal cancer cells that express truncated APC by reducing cellular cholesterol levels and inducing apoptotic cell death through the ER stress/ROS/JNK signaling in colon cancer cells. purity: 99%
Catalog No. 50-258-0780