Learn More
Medchemexpress LLC HY-14804 10mg Medchemexpress, Tasisulam CAS:519055-62-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1480410MG
Encompass_Preferred
Medchemexpress, HY-14804 10mg Tasisulam CAS:519055-62-0 Tasisulam is a anticancer agent and induces apoptosis via the intrinsic pathway, resulting in cytochrome c release and caspase-dependent cell death. Tasisulam inhibits mitotic progression and induces vascular normalization. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0556