Learn More
Medchemexpress LLC Taspoglutide | 275371-94-3 | 98.4% | 3339.71 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP01651MG
Encompass_Preferred
Taspoglutide is a long-acting glucagon-like peptide 1 (GLP-1) receptor agonist. It was developed for the treatment of type 2 diabetes and has an EC50 value of 0.06 nM. It activates the GLP-1 receptor with comparable affinity and potency to the natural ligand for the hGLP-1 receptor. Taspoglutide enhances glucose-induced insulin secretion and lowers blood glucose in models of type 2 diabetes.
  • Activates GLP-1 receptor
  • Enhances glucose-induced insulin secretion
  • Lowers blood glucose
  • Suitable for type 2 diabetes research
Catalog No. 50-004-26032