Learn More
Medchemexpress LLC Tasurgratinib succinate | 1879965-80-6 | 98.1% | 705.8 g/mol | C36H43N5O10 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101466A100MG
Encompass_Preferred
Tasurgratinib succinate is the succinate salt of tasurgratinib, a potent, orally active inhibitor selective for fibroblast growth factor receptors FGFR1, FGFR2, and FGFR3. It is used in preclinical and clinical research of FGFR-driven cancers and is supplied as a research-grade compound with supporting analytical documentation.
Catalog No. 50-002-48984
Add to Cart
Request a Quote