Learn More
Medchemexpress LLC TAT (47-57), FAM-labeled acetate | 1918.13 (free base) | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP4122A5MG
Encompass_Preferred
TAT (47-57), FAM-labeled acetate is a FAM-labeled cell-penetrating peptide (CPP) with potential for use in intracellular drug delivery studies. It is soluble in H2O at concentrations of 50 mg/mL or higher.
  • FAM-labeled cell penetrating peptide
  • Potential for use in intracellular drug delivery studies
  • Soluble in H2O
  • Used in peptide and drug delivery research
  • Fluorescent-labeled peptide
Catalog No. 50-003-98537