Learn More
Medchemexpress LLC Tat-cyclo-cllfvy | 1446322-66-2 | 98.8% | 2559.08 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP14201MG
Encompass_Preferred
TAT-cyclo-CLLFVY is a cyclic peptide inhibitor of HIF-1 heterodimerization that inhibits hypoxia signaling in cancer cells. It disrupts HIF-1α/HIF-1β protein-protein interaction with an IC50 of 1.3 μM.
  • Cyclic peptide inhibitor of HIF-1 heterodimerization
  • Inhibits hypoxia signaling in cancer cells
  • Disrupts HIF-1α/HIF-1β protein-protein interaction
  • Inhibits HIF-1 activity in mammalian cell luciferase reporter assay
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-23420