Learn More
Medchemexpress LLC TATA Box Binding Protein Antibody (YA3260) | 100 UL

Supplier: Medchemexpress LLC HYP83515100UL
TATA Box Binding Protein (TBP) antibody (Clone NO.: YA3260) is a rabbit-derived, non-conjugated IgG monoclonal antibody designed to recognize and bind promoters with or without a TATA box, facilitating the assembly of the pre-initiation complex (PIC) in RNA polymerase II (Pol II)-dependent transcription.
- Host: Rabbit
- Reactivity: Human, mouse
- Conjugation: Non-conjugated
- Applications: WB, IHC-P, IHC-F, ICC/IF, IP
- Research field: Epigenetics and nuclear signaling
- Storage conditions: Store at -20°C for 1 year; avoid repeated freeze/thaw cycles
- Purity: Affinity purified
- Molecular weight: 38 kDa (predicted and observed)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.