Learn More
Sino Biological Recombinant Human Tau Protein (full length, AVI Tag), Biotinylated
Supplier:  Sino Biological T0654B2N100
Encompass_Preferred
Recombinant full-length human Tau-410 was expressed in E. coli cells with an N-terminal Avi tag and enzymatically biotinylated in vitro prior to the final chromatography purification.
Catalog No. 50-000-83891