Learn More
Sino Biological Recombinant Human Tau Protein (AVI Tag), Biotinylated
Supplier:  Sino Biological T0855HNB20
Encompass_Preferred
Recombinant human tag-free Tau-441 (231-421) was expressed in E. coli cells with an N-terminal Avi tag and enzymatically biotinylated in vitro prior to the final chromatography purification.
Catalog No. 50-000-83875