Learn More
Enzo Life Sciences Tau (human) monoclonal antibody (C3) (1ml)
SDP
Supplier:  Enzo Life Sciences ENZABS2141000
Encompass_Preferred
Clone: C3. Isotype: IgG1. Species reactivity: Human. Specificity: Recognizes human Tau cleaved at D421. Applications: ELISA, IHC, WB. Purity: Purified. Formulation: Liquid. In PBS. Does not contain any preservatives or carrier proteins.
Catalog No. 50-201-2763