Learn More
Bachem Tau Peptide (304-318) Trifluoroacetate
SDP
Supplier:  Bachem 4099425.1000
Encompass_Preferred
Tau Peptide (304-318) Trifluoroacetate, H-Gly-Ser-Val-Gln-Ile-Val-Tyr-Lys-Pro-Val-Asp-Leu-Ser-Lys-Val-OH, 330456-49-0, C75H126N18O22, Mr 1631.93, 1mg.
Catalog No. 50-194-6639