Learn More
Cayman Chemical TauPhosphoSR396 RabbIt M 100uL
SDP
Supplier:  Cayman Chemical 35905100 UL
Encompass_Preferred
A DNA-crosslinking agent forms monofunctional DNA adducts inhibits proliferation of MDCK cells expressing OCT1 or OCT2 (IC50s 8 1 and 1 5 UM respectively) inhibits RNA polymerase II-mediated transcription in a reporter assay using HeLa cells increases survival in murine S180 sarcoma and P388 leukemia models at 40 and 80 mg/kg respectively
Catalog No. 50-290-9220