Learn More
U.S. Pharmacopeia Taurine, 107-35-7, MFCD00008197, 100mg
Supplier:  U.S. Pharmacopeia 1643361
Encompass
Molecular formula C2H7NO3S, Molecular weight 125.15, Melting point/freezing point 572 °F (300 °C) (decomposes), Initial boiling point 230 °F (110 °C) (sublimes)
Catalog No. NC1640737