Learn More
Medchemexpress LLC Taurine | 107-35-7 | 99.9% | 100 G
SDP
Supplier:  Medchemexpress LLC HYB0351100G
Encompass_Preferred
Taurine is a sulfur-containing amino acid and an organic osmolyte involved in cell volume regulation. It provides a substrate for the formation of bile salts and plays a role in modulating intracellular free calcium concentration. Taurine has the ability to activate autophagy in adipocytes.
  • Involved in cell volume regulation
  • Provides a substrate for bile salt formation
  • Modulates intracellular free calcium concentration
  • Activates autophagy in adipocytes
  • One of the most abundant amino acids in the brain and spinal cord, leukocytes, heart, muscle cells, and retina
  • Exhibits diverse biological actions including protection against ischemia-reperfusion injury
  • Offers antioxidant, antiatherogenic, and blood pressure-lowering effects
Catalog No. 50-004-07346