Learn More
Medchemexpress LLC Tauro-ω-muricholic acid sodium | 2456348-84-6 | MFCD00082254 | 97.8% | 537.68 | C26H44NNaO7S | 5 MG

Supplier: Medchemexpress LLC HY1360665MG
Tauro-ω-muricholic acid sodium is the sodium salt of a muricholic bile acid used as a research reagent in biochemical and physiological studies. It is supplied as a white to off-white solid with high reported purity and is investigated as a potential plasma marker for early-onset neonatal sepsis and cholestasis. Store and handle under recommended cold, moisture-controlled conditions.
- High purity (97.8%).
- Provided as a white to off-white solid.
- Soluble in DMSO; may require warming and ultrasonication.
- Storage stability: frozen storage recommended; in-solution time limits apply.
- Suitable for biochemical assays and analytical studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.