Learn More
Medchemexpress LLC Tauro-obeticholic acid | 863239-61-6 | MFCD33402170 | 98.2% | C28H49NO6S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13539910MG
Encompass_Preferred
Tauro-obeticholic acid is the taurine-conjugated active metabolite of obeticholic acid and functions as an agonist of the farnesoid X receptor (FXR). It is supplied for research use in studies of bile acid signaling, FXR-mediated pharmacology, and metabolic pathways; typical specifications include formula C28H49NO6S, MW 527.76, and high purity.
  • Acts as a farnesoid X receptor (FXR) agonist useful for studying bile acid signaling.
  • Taurine-conjugated metabolite of obeticholic acid.
  • Supplied with high purity suitable for analytical and pharmacology studies.
  • Available in multiple small-scale package sizes for research workflows.
  • Soluble in common organic solvents and DMSO for assay preparation.
Catalog No. 50-002-61077