Learn More
Cayman Chemical TaurocholIc AcId 3sulfATs 5mg
SDP
Supplier:  Cayman Chemical 90025385 MG
Encompass_Preferred
A metabolite of taurocholic acid; plasma levels are elevated in wild-type and Sort1 knockout mice at 6 hours post-BDL and are further elevated in Sort1 knockout mice at 24 hours post-BDL
Catalog No. 50-289-3177