Learn More
Medchemexpress LLC Taurocholic acid-d4 sodium | 2410279-93-3 | MFCD29472034 | 98.2% | 541.71 | C26H40D4NNaO7S | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB1788S10MG
Encompass_Preferred
Taurocholic acid-d4 sodium is the deuterium-labeled (d4) sodium salt of taurocholic acid supplied as a white to off-white solid. It is designed for use as an isotopically labeled internal standard or tracer in quantitative analytical methods, including NMR, LC-MS, and GC-MS, supporting accurate measurement of taurocholic acid. The material is supplied in small analytical packs and should be stored sealed, away from moisture, at recommended temperatures.
  • Isotopically labeled (d4) sodium salt for use as an internal standard.
  • Suitable for NMR, LC-MS, and GC-MS quantification.
  • High purity (>98%) with supporting documentation.
  • White to off-white solid, easily handled in analytical workflows.
  • Stable under recommended storage conditions to preserve integrity.
Catalog No. 50-002-98996