Learn More
Medchemexpress LLC Taurocholic acid-d5 sodium | 2044276-13-1 | 98.0% | 542.72 g/mol | C26H39D5NNaO7S | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW6538931MG
Encompass_Preferred
Deuterium-labeled taurocholic acid sodium (taurocholic acid-d5 sodium) is a stable isotope internal standard designed for analytical and metabolomics workflows. Supplied as a small, high-purity sodium salt powder for laboratory research use only; not for human or veterinary use.
  • Deuterium-labeled (d5) internal standard for quantitative LC-MS/MS.
  • Suitable for metabolomics and targeted bile acid analysis.
  • High chemical purity (98.0%).
  • Supplied as a 1 mg powder vial for analytical use.
  • Sodium salt form for improved aqueous solubility and handling.
Catalog No. 50-002-84720