Learn More
Medchemexpress LLC Taurocholic acid-d8 sodium | 1351996-69-4 | 98.8% | 545.73 | C26H36D8NNaO7S | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW6538421MG
Encompass_Preferred
Taurocholic acid-d8 sodium is a deuterium-labeled sodium salt of taurocholic acid supplied as an analytical standard. It is used as an internal standard for mass spectrometry, metabolic tracing, and analytical method development, offering reliable quantification through isotopic labeling.
  • Isotopically labeled (d8) sodium salt.
  • High purity: 98.83%.
  • Molecular weight 545.73.
  • Supplied in 1 mg vials.
  • Store sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-002-84425