Learn More
Cayman Chemical TaurocholIc AcIdd4sodIum s 5mg
SDP
Supplier:  Cayman Chemical 218915 MG
Encompass_Preferred
An internal standard for the quantification of taurocholic acid by GC- or LC-MS
Catalog No. 50-298-7994