Learn More
Cayman Chemical TaurocholIc AcIdodIum salt 5g
SDP
Supplier:  Cayman Chemical 162155 G
Encompass_Preferred
A taurine-conjugated form of cholic acid; serum levels of TCA are decreased in patients with Crohn’s disease and those with ulcerative colitis with no extraintestinal manifestations, but are increased in patients with ulcerative colitis accompanied by hepatobiliary disease
Catalog No. 50-297-0165