Learn More
Cayman Chemical Taurod4ursodeoxycholc ACD 10mg
SDP
Supplier:  Cayman Chemical 3985110 MG
Encompass_Preferred
An internal standard for the quantification of tauroursodeoxycholic acid by GC- or LC-MS
Catalog No. 50-294-2063