Learn More
Medchemexpress LLC Taurodeoxycholic acid sodium hydrate | 110026-03-4 | 99.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB1899A10MM/1ML
Encompass_Preferred
Taurodeoxycholic acid sodium hydrate is a bile acid and an amphiphilic surfactant molecule synthesized from cholesterol in the liver. It activates the S1PR2 pathway in addition to the TGR5 pathway.
  • Bile acid
  • Amphiphilic surfactant molecule
  • Synthesized from cholesterol in the liver
  • Activates S1PR2 pathway
  • Activates TGR5 pathway
  • Appears as a white to off-white solid
Catalog No. 50-004-12287