Learn More
Medchemexpress LLC Taurohyocholic acid | 32747-07-2 | 99.7% | 1 MG

Supplier: Medchemexpress LLC HY1217991MG
Taurohyocholic acid is a taurine-conjugated hyocholic bile acid used as an endogenous metabolite standard and research reagent in studies of bile acid metabolism and pharmacology.
- Provided as a high-purity analytical standard (99.7%).
- Molecular weight 515.70 g/mol; formula C26H45NO7S.
- Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
- Powder stable at -20°C for up to three years; solutions stable at -80°C for months.
- Available in small milligram pack sizes suitable for analytical and reference use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.